Gene Transcripts

Gene Symbol
Cftr
Entrez Gene ID
12638
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
AW495489, Abcc7
Chromosome Location
Chr. 6: 18170262 - 18322774 on Build GRCm38
UniGene ID
Mm.15621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021050.2 NP_066388.1 23 3937
GenBank mRNA M60493.1 - 23 3938
M69298.1 - 23 3936

View Full Product Details