Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
14067
Gene Name
coagulation factor V
Gene Aliases
AI173222, Cf-5, Cf5
Chromosome Location
Chr. 1: 164151859 - 164220274 on Build GRCm38
UniGene ID
Mm.12900
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK137521.1 - 13 2805
AK137537.1 - 13 2782
AK139240.1 - 13 2799
U52925.1 - 13 2680

View Full Product Details