Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175088.5 | NP_780297.3 | 5 | 1107 | |
GenBank mRNA | AK048821.1 | - | 5 | 1047 | |
AK086117.1 | - | 6 | 1110 | ||
AK165016.1 | - | 5 | 1108 | ||
AK172522.1 | - | 4 | 956 | ||
BC067006.1 | - | 5 | 838 |