Gene Transcripts
- Gene Symbol
- Rab1
- Entrez Gene ID
- 19324
- Gene Name
- RAB1, member RAS oncogene family
- Gene Aliases
- Gtbp, Rab-1, Rab1A, Ypt1, mKIAA3012
- Chromosome Location
- Chr. 11: 20201602 - 20226856 on Build GRCm38
- UniGene ID
- Mm.271944
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008996.3 |
NP_033022.1
|
5, 6 |
458 |
|
GenBank mRNA |
AB232583.1 |
- |
5, 6 |
430 |
|
AF226873.1 |
- |
5, 6 |
458 |
|
AK034408.1 |
- |
5, 6 |
626 |
|
AK129477.1 |
- |
5, 6 |
616 |
|
AK151085.1 |
- |
5, 6 |
622 |
|
AK152978.1 |
- |
5, 6 |
627 |
|
AK159425.1 |
- |
5, 6 |
622 |
|
AK166249.1 |
- |
5, 6 |
619 |
|
AK167150.1 |
- |
5, 6 |
626 |
|
AK207850.1 |
- |
1 |
33 |
|
AK212539.1 |
- |
1, 2 |
128 |
|
AK212657.1 |
- |
1, 2 |
149 |
|
BC002077.1 |
- |
5, 6 |
625 |
3592802 |
Y00094.1 |
- |
5, 6 |
559 |
|
View Full Product Details