Gene Transcripts

Gene Symbol
Rangap1
Entrez Gene ID
19387
Gene Name
RAN GTPase activating protein 1
Gene Aliases
C79654, Fug1, mKIAA1835
Chromosome Location
Chr. 15: 81704248 - 81729919 on Build GRCm38
UniGene ID
Mm.270975
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146174.1 NP_001139646.1 2, 3 356
NM_011241.4 NP_035371.4 3, 4 404
GenBank mRNA AK035571.1 - 3, 4 366
AK042930.1 - 2, 3 321
AK088768.1 - 2, 3 278
AK129452.1 - 2, 3 285
AK133905.1 - 2, 3 321
AK145459.1 - 2, 3 277
AK146670.1 - 3, 4 364
AK159009.1 - 2, 3 309
AK159525.1 - 2, 3 321
AK159930.1 - 2, 3 312
AK181740.1 - 1, 2 35
BC014855.1 - 2, 3 312 3157225
BC052862.1 - 2, 3 279 30063509
BC066213.1 - 2, 3 320 30466595
BC071200.1 - 3, 4 520 5032610
U08110.1 - 3, 4 387

View Full Product Details