Gene Transcripts
- Gene Symbol
- Sepp1
- Entrez Gene ID
- 20363
- Gene Name
- selenoprotein P, plasma, 1
- Gene Aliases
- AU018766, D15Ucla1, Se-P, selp
- Chromosome Location
- Chr. 15: 3270767 - 3280508 on Build GRCm38
- UniGene ID
- Mm.392203
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042613.1 |
NP_001036078.1
|
2, 3 |
282 |
|
NM_001042614.1 |
NP_001036079.1
|
2, 3 |
365 |
|
NM_009155.3 |
NP_033181.3
|
2, 3 |
299 |
|
GenBank mRNA |
AI892015.1 |
- |
2, 3 |
374 |
|
AK002450.1 |
- |
2, 3 |
281 |
|
AK077265.1 |
- |
2, 3 |
281 |
|
AK146774.1 |
- |
2, 3 |
281 |
|
AK148623.1 |
- |
2, 3 |
281 |
|
AK159269.1 |
- |
2, 3 |
281 |
|
AK159524.1 |
- |
2, 3 |
282 |
|
AK159894.1 |
- |
2, 3 |
278 |
|
AK159911.1 |
- |
2, 3 |
282 |
|
AK159935.1 |
- |
2, 3 |
281 |
|
AK167611.1 |
- |
2, 3 |
281 |
|
AK168463.1 |
- |
2, 3 |
278 |
|
AK168583.1 |
- |
2, 3 |
278 |
|
AK168631.1 |
- |
2, 3 |
281 |
|
AK168747.1 |
- |
2, 3 |
281 |
|
AK168926.1 |
- |
2, 3 |
281 |
|
AK169011.1 |
- |
2, 3 |
280 |
|
AK169158.1 |
- |
2, 3 |
280 |
|
AK169362.1 |
- |
2, 3 |
281 |
|
AK190911.1 |
- |
1, 2 |
116 |
|
AK196001.1 |
- |
1, 2 |
116 |
|
AK200678.1 |
- |
1, 2 |
116 |
|
AK201439.1 |
- |
1, 2 |
117 |
|
AK201752.1 |
- |
1, 2 |
116 |
|
AK203751.1 |
- |
1, 2 |
116 |
|
BC001991.1 |
- |
2, 3 |
282 |
3486761 |
CB231357.1 |
- |
2, 3 |
302 |
|
CK627189.1 |
- |
2, 3 |
282 |
|
X99807.1 |
- |
1, 2 |
342 |
|
View Full Product Details