Gene Transcripts

Gene Symbol
Slc22a2
Entrez Gene ID
20518
Gene Name
solute carrier family 22 (organic cation transporter), member 2
Gene Aliases
Oct2, Orct2
Chromosome Location
Chr. 17: 12583958 - 12628488 on Build GRCm38
UniGene ID
Mm.17322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013667.2 NP_038695.1 1 110
GenBank mRNA AJ006036.1 - 1 110
AK052658.1 - 1 107
AK143892.1 - 1 109
BC015250.1 - 1 38 4242038
BC069911.1 - 1 57 4974824

View Full Product Details