Gene Transcripts

Gene Symbol
Tigit
Entrez Gene ID
100043314
Gene Name
T cell immunoreceptor with Ig and ITIM domains
Gene Aliases
ENSMUSG00000071552, Vstm3
Chromosome Location
Chr. 16: 43648861 - 43664184 on Build GRCm38
UniGene ID
Mm.390896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146325.1 NP_001139797.1 2 293
GenBank mRNA EU675311.1 - 2 255

View Full Product Details