Gene Transcripts

Gene Symbol
Slc9c1
Entrez Gene ID
208169
Gene Name
solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1
Gene Aliases
Gm610, NHE-10, Slc9a10, sNHE, spermNHE
Chromosome Location
Chr. 16: 45535266 - 45610396 on Build GRCm38
UniGene ID
Mm.277437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198106.4 NP_932774.3 16 2088
GenBank mRNA AY368685.1 - 16 2048
BC145031.1 - 1695
BC145032.1 - 1909

View Full Product Details