Gene Transcripts

Gene Symbol
Nek11
Entrez Gene ID
208583
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 11
Gene Aliases
4932416N14Rik
Chromosome Location
Chr. 9: 105162156 - 105395524 on Build GRCm38
UniGene ID
Mm.347509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172461.3 NP_766049.2 16 1977
GenBank mRNA AK030042.1 - 16 1936
BC119588.1 - 15 1811 40058889
BC119589.1 - 15 1811 40058890

View Full Product Details