Gene Transcripts

Gene Symbol
Plekhg1
Entrez Gene ID
213783
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 1
Gene Aliases
D10Ertd733e, Gm521, mKIAA1209
Chromosome Location
Chr. 10: 3740251 - 3967303 on Build GRCm38
UniGene ID
Mm.150831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033253.3 NP_001028425.1 9 1309
NM_001159942.1 NP_001153414.1 10 1475
GenBank mRNA AK122468.1 - 6 633
AK135923.1 - 9 1311
BC076628.1 - 7 706
BC117965.1 - 8 911 40098326
BC117966.1 - 8 904 40098330

View Full Product Details