Gene Transcripts

Gene Symbol
Zfp1
Entrez Gene ID
22640
Gene Name
zinc finger protein 1
Gene Aliases
Fnp-1, Zfp-1, mkr-1
Chromosome Location
Chr. 8: 111643443 - 111671108 on Build GRCm38
UniGene ID
Mm.4184
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037665.2 NP_001032754.1 4 669
NM_011742.2 NP_035872.2 4 627
GenBank mRNA AK134452.1 - 4 534
AK147054.1 - 4 607
AK153830.1 - 4 611
BC025141.1 - 5 739
BC055796.1 - 4 578 6412519
BC062970.1 - 4 578 6412519
X16493.1 - 4 736

View Full Product Details