Gene Transcripts

Gene Symbol
Ralgapb
Entrez Gene ID
228850
Gene Name
Ral GTPase activating protein, beta subunit (non-catalytic)
Gene Aliases
9330195P16, AI507211, B230339M05Rik, mKIAA1219, p170
Chromosome Location
Chr. 2: 158409829 - 158499253 on Build GRCm38
UniGene ID
Mm.398185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291137.1 NP_001278066.1 19, 20 3020
NM_001291138.1 NP_001278067.1 18, 19 2984
GenBank mRNA AK122470.1 - 14, 15 1734
BC138377.1 - 19, 20 2922 8861397
BC145260.1 - 18, 19 2886
CB844140.2 - 1, 2 135
CN458381.1 - 4, 5 592

View Full Product Details