Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026079.3 | NP_080355.2 | 19 | 2304 | |
GenBank mRNA | AF140786.1 | - | 19 | 2232 | |
AF367244.1 | - | 19 | 2257 | ||
AF387811.1 | - | 19 | 2271 | ||
AK031367.1 | - | 19 | 2249 | ||
AK035944.1 | - | 19 | 2306 | ||
AK147153.1 | - | 19 | 2307 | ||
AK165168.1 | - | 16 | 1781 | ||
BC052387.1 | - | 19 | 2216 | 5705589 |