Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146150.2 | NP_666262.2 | 30, 31 | 3317 | |
GenBank mRNA | AK031548.1 | - | 30, 31 | 3317 | |
AK050944.1 | - | 15, 16 | 1416 | ||
AK166470.1 | - | 28, 29 | 3316 | ||
BC023786.1 | - | 29, 30 | 2695 | ||
BC024544.1 | - | 14, 15 | 1302 | ||
BC026832.1 | - | 30, 31 | 3367 | 4486176 | |
BC029977.1 | - | 21, 22 | 1953 | ||
BC036128.1 | - | 30, 31 | 3435 | 4971791 |