Gene Transcripts

Gene Symbol
Vmn2r66
Entrez Gene ID
233437
Gene Name
vomeronasal 2, receptor 66
Gene Aliases
F830104D24Rik
Chromosome Location
Chr. 7: 84994645 - 85012020 on Build GRCm38
UniGene ID
Mm.424396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033878.3 NP_001029050.3 2 455

View Full Product Details