Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252520.1 | NP_001239449.1 | 1 | 405 | |
NM_001252521.1 | NP_001239450.1 | 1 | 405 | ||
NM_001252522.1 | NP_001239451.1 | 1 | 405 | ||
NM_001252523.1 | NP_001239452.1 | 1 | 405 | ||
NM_001252524.1 | NP_001239453.1 | 1 | 405 | ||
NM_146194.4 | NP_666306.2 | 1 | 405 | ||
GenBank mRNA | AK144960.1 | - | 1 | 143 | |
AK162360.1 | - | 1 | 98 | ||
AK170373.1 | - | 1 | 425 | ||
AK171915.1 | - | 1 | 142 | ||
AK220291.1 | - | 1 | 121 | ||
AY206701.1 | - | 1 | 89 | ||
BC011470.1 | - | 1 | 115 | 2651109 | |
BC021491.1 | - | 1 | 354 | 5345558 | |
BC023843.1 | - | 1 | 153 | 5346041 | |
BC025566.1 | - | 1 | 428 | 5322606 | |
BC057683.1 | - | 1 | 356 | 3671962 |