Gene Transcripts

Gene Symbol
Ogdhl
Entrez Gene ID
239017
Gene Name
oxoglutarate dehydrogenase-like
Gene Aliases
Chromosome Location
Chr. 14: 32322012 - 32348150 on Build GRCm38
UniGene ID
Mm.318302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081130.1 NP_001074599.1 3, 4 373
GenBank mRNA BC157970.1 - 2, 3 230
BC157971.1 - 2, 3 230 9007293


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DV658608.1 - 247 Chr. - - [-]

View Full Product Details