Gene Transcripts

Gene Symbol
Tspyl5
Entrez Gene ID
239364
Gene Name
testis-specific protein, Y-encoded-like 5
Gene Aliases
6330530B20, E130308C19Rik
Chromosome Location
Chr. 15: 33683875 - 33687883 on Build GRCm38
UniGene ID
Mm.60794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085421.1 NP_001078890.1 1 740
GenBank mRNA AK031988.1 - 1 451
AK087510.1 - 1 676
AK173253.1 - 1 683
BC079547.1 - 1 691

View Full Product Details