Gene Transcripts

Gene Symbol
Mab21l2
Entrez Gene ID
23937
Gene Name
mab-21-like 2 (C. elegans)
Gene Aliases
Chromosome Location
Chr. 3: 86545581 - 86548283 on Build GRCm38
UniGene ID
Mm.389466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011839.3 NP_035969.2 1 754
GenBank mRNA AF126427.1 - 1 1095
AF149877.1 - 1 496
AK053645.1 - 1 755
AK144373.1 - 1 923
BC046628.1 - 1 720 6493460
BC058730.1 - 1 860

View Full Product Details