Gene Transcripts

Gene Symbol
Sh3bp2
Entrez Gene ID
24055
Gene Name
SH3-domain binding protein 2
Gene Aliases
3BP2
Chromosome Location
Chr. 5: 34525784 - 34563639 on Build GRCm38
UniGene ID
Mm.5012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145858.1 NP_001139330.1 9, 10 1566
NM_001145859.1 NP_001139331.1 9, 10 1569
NM_011893.3 NP_036023.3 9, 10 1466
GenBank mRNA AK089455.1 - 9, 10 1397
AK143548.1 - 9, 10 1571
AK150265.1 - 9, 10 1568
AK155103.1 - 9, 10 1423
AK155898.1 - 9, 10 1466
AK155970.1 - 9, 10 1514
BC010198.1 - 9, 10 1404 3967543
BC085497.1 - 9, 10 1457 5696296
L14543.1 - 9, 10 1400

View Full Product Details