Gene Transcripts

Gene Symbol
Fbxo46
Entrez Gene ID
243867
Gene Name
F-box protein 46
Gene Aliases
20D7-FC4, 4932704E22Rik, Fbxo34l
Chromosome Location
Chr. 7: 19119859 - 19138261 on Build GRCm38
UniGene ID
Mm.230830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175530.3 NP_780739.1 2 1260
GenBank mRNA AK030147.1 - 2 1249
AK030220.1 - 2 1246
BC049813.1 - 2 1234 6507415

View Full Product Details