Gene Transcripts
- Gene Symbol
- Lgi4
- Entrez Gene ID
- 243914
- Gene Name
- leucine-rich repeat LGI family, member 4
- Gene Aliases
- Lgil3, clp
- Chromosome Location
- Chr. 7: 31058953 - 31070935 on Build GRCm38
- UniGene ID
- Mm.389870
- Species
- Mus musculus
- Gene Symbol
- Fxyd3
- Entrez Gene ID
- 17178
- Gene Name
- FXYD domain-containing ion transport regulator 3
- Gene Aliases
- Mat8
- Chromosome Location
- Chr. 7: 31070535 - 31076697 on Build GRCm38
- UniGene ID
- Mm.263847
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK145206.1 |
- |
|
2090 |
|
View Full Product Details