Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153591.4 | NP_705819.3 | 11 | 1409 | |
GenBank mRNA | AK046799.1 | - | 11 | 1410 | |
AK046801.1 | - | 11 | 1410 | ||
AK047091.1 | - | 11 | 1354 | ||
AK050718.1 | - | 11 | 1348 | ||
AK138260.1 | - | 11 | 1389 | ||
AK164179.1 | - | 11 | 1378 | ||
AK188846.1 | - | 5 | 352 | ||
AK209841.1 | - | 4 | 284 | ||
BC030041.1 | - | 7 | 613 | 1226297 |