Gene Transcripts

Gene Symbol
Klhl34
Entrez Gene ID
245683
Gene Name
kelch-like 34
Gene Aliases
Gm388
Chromosome Location
Chr. X: 157818435 - 157821060 on Build GRCm38
UniGene ID
Mm.295846
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081667.2 NP_001075136.1 1 13

View Full Product Details