Gene Transcripts

Gene Symbol
Eif2s3y
Entrez Gene ID
26908
Gene Name
eukaryotic translation initiation factor 2, subunit 3, structural gene Y-linked
Gene Aliases
Eif-2gy, Spy, Tfy
Chromosome Location
Chr. Y: 1010578 - 1046811 on Build GRCm38
UniGene ID
Mm.250909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012011.1 NP_036141.1 12 1384
GenBank mRNA AJ006584.1 - 12 1384
AK009448.1 - 12 1389
AK153974.1 - 12 1396
BC031450.1 - 10 2459
BC043656.1 - 12 1384 5368023

View Full Product Details