Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138584.2 | NP_613050.1 | 9 | 1083 | |
GenBank mRNA | AK009332.1 | - | 9 | 1075 | |
AK011985.1 | - | 9 | 1050 | ||
AK150215.1 | - | 9 | 1084 | ||
AK150581.1 | - | 9 | 1061 | ||
AK153284.1 | - | 9 | 1045 | ||
AK170666.1 | - | 9 | 1076 | ||
AK219439.1 | - | 2 | 106 | ||
BC099434.1 | - | 8 | 848 | 30714184 | |
BC132377.1 | - | 9 | 1060 | 40130654 | |
BC132403.1 | - | 9 | 1060 | 40130680 |