Gene Transcripts

Gene Symbol
Pttg1
Entrez Gene ID
30939
Gene Name
pituitary tumor-transforming gene 1
Gene Aliases
AW555095, C87862, Pttg, Pttg3
Chromosome Location
Chr. 11: 43420248 - 43426248 on Build GRCm38
UniGene ID
Mm.6856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013917.2 NP_038945.2 4 821
GenBank mRNA AF071209.1 - 4 821

View Full Product Details