Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177016.3 | NP_795990.2 | 4 | 526 | |
GenBank mRNA | AK033669.1 | - | 4 | 529 | |
AK050272.1 | - | 4 | 530 | ||
AK162320.1 | - | 4 | 525 | ||
BC147415.1 | - | 4 | 487 | 9056491 | |
BC147416.1 | - | 4 | 487 | 9056498 |