Gene Transcripts

Gene Symbol
Lingo4
Entrez Gene ID
320747
Gene Name
leucine rich repeat and Ig domain containing 4
Gene Aliases
A530050P17Rik, LERN4, Lrrn6d
Chromosome Location
Chr. 3: 94399219 - 94404501 on Build GRCm38
UniGene ID
Mm.213001
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177250.2 NP_796224.1 2 1116
GenBank mRNA AK040952.1 - 2 1116
AK042286.1 - 1817
BC117866.1 - 2 1026 40095465
BC117867.1 - 2 1026 40095468

View Full Product Details