Gene Transcripts

Gene Symbol
Ltv1
Entrez Gene ID
353258
Gene Name
LTV1 homolog (S. cerevisiae)
Gene Aliases
2610020N02Rik
Chromosome Location
Chr. 10: 13178597 - 13193137 on Build GRCm38
UniGene ID
Mm.117581
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181470.4 NP_852135.1 2 225
GenBank mRNA AK011491.1 - 2 211
BC067062.1 - 2 225 30538957
BC069962.1 - 2 219 5038459

View Full Product Details