Gene Transcripts

Gene Symbol
Abca17
Entrez Gene ID
381072
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 17
Gene Aliases
Chromosome Location
Chr. 17: 24264258 - 24351897 on Build GRCm38
UniGene ID
Mm.328842
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031621.2 NP_001026792.2 2 241
GenBank mRNA AB112584.1 - 2 241
DQ660313.1 - 3 336

View Full Product Details