Gene Transcripts

Gene Symbol
Pnma5
Entrez Gene ID
385377
Gene Name
paraneoplastic antigen family 5
Gene Aliases
mKIAA1934
Chromosome Location
Chr. X: 73033981 - 73040203 on Build GRCm38
UniGene ID
Mm.327861
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100461.3 NP_001093931.1 1 816
GenBank mRNA AK220432.1 - 1 816

View Full Product Details