Gene Transcripts

Gene Symbol
Hs6st3
Entrez Gene ID
50787
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
6OST3, HS6ST-3
Chromosome Location
Chr. 14: 119137660 - 119872871 on Build GRCm38
UniGene ID
Mm.445777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015820.3 NP_056635.2 2 1402
GenBank mRNA AB024567.1 - 2 1365
BC117529.1 - 3 1220 40106840
BC118035.1 - 2 1326 40106835

View Full Product Details