Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290536.1 | NP_001277465.1 | 6 | 1442 | |
NM_001290537.1 | NP_001277466.1 | 7 | 1062 | ||
NM_138604.3 | NP_613070.2 | 6 | 1442 | ||
GenBank mRNA | AF229642.1 | - | 6 | 583 | |
AK010750.1 | - | 7 | 1062 | ||
AK149777.1 | - | 7 | 1063 | ||
AK155131.1 | - | 6 | 1418 | ||
AK167226.1 | - | 5 | 755 | ||
AK169921.1 | - | 8 | 1028 | ||
AK171874.1 | - | 6 | 1183 | ||
BC016529.1 | - | 6 | 1442 | 3594583 | |
BC051111.1 | - | 6 | 1181 | 5253679 |