Gene Transcripts

Gene Symbol
Trim3
Entrez Gene ID
55992
Gene Name
tripartite motif-containing 3
Gene Aliases
BERP1, HAC1, Rnf22
Chromosome Location
Chr. 7: 105604463 - 105633571 on Build GRCm38
UniGene ID
Mm.29290
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285870.1 NP_001272799.1 10 2156
NM_001285871.1 NP_001272800.1 10 2341
NM_001285873.1 NP_001272802.1 10 2156
NM_018880.3 NP_061368.1 10 2130
GenBank mRNA AB030912.1 - 10 2157
AF220019.1 - 10 2049
AK019165.1 - 10 2091
AK144674.1 - 10 2059
AK170029.1 - 10 2094
AK182206.1 - 1 69
BC003347.1 - 10 2170
BC034263.1 - 10 2095 4503884

View Full Product Details