Gene Transcripts

Gene Symbol
Shank3
Entrez Gene ID
58234
Gene Name
SH3/ankyrin domain gene 3
Gene Aliases
AI841104, SHANK3c-3, SHANK3c-4, Shank3b
Chromosome Location
Chr. 15: 89499707 - 89560261 on Build GRCm38
UniGene ID
Mm.146855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021423.3 NP_067398.2 19, 20 2257
GenBank mRNA AB231013.1 - 19, 20 2257
AB841411.1 - 9, 10 1221
AB841412.1 - 9, 10 1221
AJ245904.1 - 16, 17 2087
KF612593.1 - 2, 3 391

View Full Product Details