Gene Transcripts

Gene Symbol
Slmo2
Entrez Gene ID
66390
Gene Name
slowmo homolog 2 (Drosophila)
Gene Aliases
2310042G06Rik, AA407013
Chromosome Location
Chr. 2: 174465076 - 174472991 on Build GRCm38
UniGene ID
Mm.182294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025531.2 NP_079807.1 6 610
GenBank mRNA AK003043.1 - 6 584
AK008542.1 - 6 570
AK013197.1 - 6 577
AK150668.1 - 6 586
BC004775.1 - 5 408
BC026968.1 - 6 610 4235395

View Full Product Details