Gene Transcripts
- Gene Symbol
- Psmd5
- Entrez Gene ID
- 66998
- Gene Name
- proteasome (prosome, macropain) 26S subunit, non-ATPase, 5
- Gene Aliases
- 1500032A03Rik, AW475925, S5b, W91691, mKIAA0072
- Chromosome Location
- Chr. 2: 34852089 - 34870962 on Build GRCm38
- UniGene ID
- Mm.270632
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080554.2 |
NP_542121.2
|
8, 9 |
1147 |
|
GenBank mRNA |
AK005335.1 |
- |
8, 9 |
1125 |
|
AK011026.1 |
- |
8, 9 |
1124 |
|
AK029067.1 |
- |
6, 7 |
2524 |
|
AK032992.1 |
- |
8, 9 |
1125 |
|
AK078419.1 |
- |
8, 9 |
1125 |
|
AK145587.1 |
- |
8, 9 |
1124 |
|
AK148284.1 |
- |
|
1148 |
|
AK210792.1 |
- |
1, 2 |
87 |
|
AK220321.1 |
- |
8, 9 |
1088 |
|
BC010650.1 |
- |
2, 3 |
248 |
|
BC019112.1 |
- |
8, 9 |
1158 |
|
View Full Product Details