Gene Transcripts

Gene Symbol
Tomm34
Entrez Gene ID
67145
Gene Name
translocase of outer mitochondrial membrane 34
Gene Aliases
2610100K07Rik, TOM34
Chromosome Location
Chr. 2: 164053538 - 164071177 on Build GRCm38
UniGene ID
Mm.23173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291155.1 NP_001278084.1 3 442
NM_025996.5 NP_080272.1 3 821
GenBank mRNA AK011786.1 - 3 821
AK017699.1 - 3 387
AK075873.1 - 3 439
AK144450.1 - 1 756
AK168329.1 - 3 376
BC018278.1 - 3 372 4187356
CX235437.1 - 3 361

View Full Product Details