Gene Transcripts

Gene Symbol
Prl7c1
Entrez Gene ID
67505
Gene Name
prolactin family 7, subfamily c, member 1
Gene Aliases
1600017N11Rik, PLP-O, Prlpo, Prplo
Chromosome Location
Chr. 13: 27773495 - 27780843 on Build GRCm38
UniGene ID
Mm.49048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026206.2 NP_080482.1 2 140
GenBank mRNA AF525157.1 - 2 125
AK005486.1 - 2 141
AK014436.1 - 2 124
BC099442.1 - 2 100 30716548

View Full Product Details