Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030241.2 | NP_084517.2 | 7 | 794 | |
GenBank mRNA | AK010593.1 | - | 6 | 793 | |
AK010835.1 | - | 6 | 819 | ||
AK030904.1 | - | 7 | 788 | ||
AK088097.1 | - | 6 | 641 | ||
AK201614.1 | - | 2 | 89 | ||
BC108333.1 | - | 7 | 723 | 4978041 |