Gene Transcripts
- Gene Symbol
- Mcoln2
- Entrez Gene ID
- 68279
- Gene Name
- mucolipin 2
- Gene Aliases
- 3300002C04Rik, AI549968, C86638, TRPML2
- Chromosome Location
- Chr. 3: 146149796 - 146195514 on Build GRCm38
- UniGene ID
- Mm.116862
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005846.2 |
NP_001005846.1
|
8, 9 |
1030 |
|
NM_026656.4 |
NP_080932.2
|
8, 9 |
1058 |
|
GenBank mRNA |
AF503575.1 |
- |
8, 9 |
1047 |
|
AK014467.1 |
- |
8, 9 |
961 |
|
AK019454.1 |
- |
8, 9 |
927 |
|
AK150446.1 |
- |
8, 9 |
1026 |
|
AK150547.1 |
- |
8, 9 |
1026 |
|
AK152067.1 |
- |
8, 9 |
1026 |
|
AK152943.1 |
- |
8, 9 |
1026 |
|
AK153290.1 |
- |
8, 9 |
1026 |
|
AK159172.1 |
- |
8, 9 |
1058 |
|
AY083532.1 |
- |
7, 8 |
876 |
|
BC029847.1 |
- |
8, 9 |
907 |
3967456 |
View Full Product Details