Gene Transcripts

Gene Symbol
Rere
Entrez Gene ID
68703
Gene Name
arginine glutamic acid dipeptide (RE) repeats
Gene Aliases
1110033A15Rik, AI414665, ARG, ARP, ATN1L, AW742570, Atr2, DNB1, eye<m03Jus>, eyes3, mKIAA0458
Chromosome Location
Chr. 4: 150281549 - 150621966 on Build GRCm38
UniGene ID
Mm.291274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085492.1 NP_001078961.1 18 3053
GenBank mRNA AK122287.1 - 1 735

View Full Product Details