Gene Transcripts

Gene Symbol
Lrrcc1
Entrez Gene ID
71710
Gene Name
leucine rich repeat and coiled-coil domain containing 1
Gene Aliases
1200008A14Rik, 4932441F23Rik, AI195358, AI447421
Chromosome Location
Chr. 3: 14533788 - 14572658 on Build GRCm38
UniGene ID
Mm.257449
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163579.1 NP_001157051.1 7 1150
NM_001163580.1 NP_001157052.1 7 1102
NM_028915.3 NP_083191.2 7 1150
GenBank mRNA AK004646.1 - 7 1151
AK016559.1 - 7 1089
AK029563.1 - 7 1067
AK173256.1 - 7 1078
BC061202.1 - 7 1058
BC099680.1 - 7 1066
BC138734.1 - 7 1124 8861756
BC138735.1 - 7 1124 8861757

View Full Product Details