Gene Transcripts

Gene Symbol
Plxdc1
Entrez Gene ID
72324
Gene Name
plexin domain containing 1
Gene Aliases
2410003I07Rik, AI848450, Tem7
Chromosome Location
Chr. 11: 97923237 - 97986667 on Build GRCm38
UniGene ID
Mm.39617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163608.1 NP_001157080.1 9 1095
NM_028199.3 NP_082475.3 9 1074
GenBank mRNA AF378760.1 - 9 1008
AK010361.1 - 9 1074
AK036144.1 - 9 1049
AK040620.1 - 8 951
AK153684.1 - 9 1061
BC114352.2 - 9 994 40057410

View Full Product Details