Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028773.3 | NP_083049.1 | 5, 6 | 735 | |
GenBank mRNA | AJ306422.1 | - | 5, 6 | 601 | |
AK004734.2 | - | 5, 6 | 736 | ||
AK156202.1 | - | 5, 6 | 800 | ||
AK171017.1 | - | 5, 6 | 733 | ||
BC028773.1 | - | 2, 3 | 294 | ||
BC064734.1 | - | 5, 6 | 720 |