Gene Transcripts

Gene Symbol
Acsl3
Entrez Gene ID
74205
Gene Name
acyl-CoA synthetase long-chain family member 3
Gene Aliases
2610510B12Rik, Acs3, C85929, Facl3, Pro2194
Chromosome Location
Chr. 1: 78657825 - 78707743 on Build GRCm38
UniGene ID
Mm.276016
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033606.2 NP_001028778.2 14 2219
NM_001136222.1 NP_001129694.1 13 1801
NM_028817.3 NP_083093.2 15 2359
GenBank mRNA AK012088.1 - 14 2162
AK160839.1 - 13 1745
AK161708.1 - 15 2360
AK166723.1 - 15 2298
BC031529.1 - 13 1944 4009004

View Full Product Details