Gene Transcripts

Gene Symbol
Spaca3
Entrez Gene ID
75622
Gene Name
sperm acrosome associated 3
Gene Aliases
1700025M08Rik, ALLP17, Lyc3, SLLP1, mSLLP1
Chromosome Location
Chr. 11: 80858389 - 80867814 on Build GRCm38
UniGene ID
Mm.26988
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029367.1 NP_083643.1 4 583
GenBank mRNA AK006357.1 - 5 731
AY572448.1 - 4 546
AY601763.1 - 3 266
BC100503.1 - 4 583 30917230

View Full Product Details