Gene Transcripts
- Gene Symbol
- Pik3r4
- Entrez Gene ID
- 75669
- Gene Name
- phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 4, p150
- Gene Aliases
- 2210010O15Rik, C730038E05Rik, C85833, D9Ertd418e
- Chromosome Location
- Chr. 9: 105642995 - 105687655 on Build GRCm38
- UniGene ID
- Mm.274830
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081309.1 |
NP_001074778.1
|
2, 3 |
1180 |
|
GenBank mRNA |
AK042361.1 |
- |
2, 3 |
975 |
|
AK042953.1 |
- |
2, 3 |
1168 |
|
AK050333.1 |
- |
2, 3 |
1150 |
|
AK054559.1 |
- |
2, 3 |
1197 |
|
AK087938.1 |
- |
2, 3 |
1171 |
|
AK169508.1 |
- |
1, 2 |
1967 |
|
BC017537.1 |
- |
1, 2 |
782 |
|
BC157947.1 |
- |
2, 3 |
1005 |
9007281 |
BC158047.1 |
- |
2, 3 |
1005 |
9088160 |
View Full Product Details